missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (CL15572) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
308.00 € - 561.00 €
Spezifikation
| Antigen | CD38 |
|---|---|
| Klon | CL15572 |
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 1 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30279678
|
Novus Biologicals
NBP3-43674-25ul |
25 μL |
308.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30279445
|
Novus Biologicals
NBP3-43674-100ul |
100 μL |
561.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Spezifikation
| CD38 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 952 | |
| IgG1 | |
| Protein A purified |
| CL15572 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| This antibody was generated using a recombinant protein sequence of P28907, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts