missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD43/Sialophorin Overexpression Lysate

Artikelnummer. 18776493 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0,1 mg
Unit Size:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
18776493 0,1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18776493 Supplier Novus Biologicals™ Supplier No. NBL116412
 Eingeschränkter Artikel
Verfügbar ab: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The CD43/Sialophorin Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Zelllinie The lysate was created in HEK293T cells, using plasmid ID RC24195 and based on accession number NM_3123. The protein contains a C-terminal DDK tag.
Zur Verwendung mit (Anwendung) Western Blot
Menge 0,1 mg
Lagerungspuffer RIPA buffer
Reinheit Protein
Testspezifität Homo sapiens sialophorin (SPN), transcript variant 2, mRNA.
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Protein State Native

Nur für Forschungszwecke

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.