missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Antibody (2B11 + PD7/26) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
689.00 € - 4140.00 €
Spezifikation
| Antigen | CD45 |
|---|---|
| Klon | 2B11 + PD7/26 |
| Konzentration | 1.0 mg/mL |
| Verdünnung | Western Blot 0.5-1.0ug/mL, Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry -frozen 0.5-1.0ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL, CyTOF-ready |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18432422
|
Novus Biologicals
NBP2-34528-1mg |
1 mg | |||||||
|
18438941
|
Novus Biologicals
NBP2-34528-0.1mg |
0.1 mg |
689.00 €
0.10 Milligramm |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
CD45 Monoclonal antibody specifically detects CD45 in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOFSpezifikation
| CD45 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 10 mM PBS | |
| 5788 | |
| IgG1 κ | |
| Protein A or G purified |
| 2B11 + PD7/26 | |
| Western Blot 0.5-1.0ug/mL, Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry -frozen 0.5-1.0ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Canine | |
| B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen | |
| Isolated neoplastic cells from T cell lymphoma (2B11); and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26) | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts