Learn More
Invitrogen™ CD45R (B220) Monoclonal Antibody (RA3-6B2), NovaFluor™ Violet 745, eBioscience™
Rat Monoclonal Antibody
188.00 € - 362.00 €
Spezifikation
| Antigen | CD45R (B220) |
|---|---|
| Klon | RA3-6B2 |
| Konzentration | 0.1 mg/mL |
| Inhalt und Lagerung | 4°C, store in dark, DO NOT FREEZE! |
| Anwendungen | Flow Cytometry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30282900
|
Invitrogen™
M011T02V02-A |
25 μg |
188.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30282585
|
Invitrogen™
M011T03V02-A |
100 μg |
362.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen. This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer. Applications Tested: This RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest Master mixes.
CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.Spezifikation
| CD45R (B220) | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| NovaFluor Violet 745 | |
| Rat | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P06800, P08575 | |
| 19264, 5788 | |
| Primary | |
| Affinity chromatography |
| RA3-6B2 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human, Mouse | |
| PTPRC | |
| B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| PTPRC | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.