missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RC Antibody (OX-22), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-43540
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD45RC Monoclonal antibody specifically detects CD45RC in Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation
Spezifikation
| CD45RC | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| OX-22 was prepared by immunizing BALB/c mice with phytohemagglutinin-activated rat lymph node cells. (Uniprot# P04157) | |
| 0.2 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| OX-22 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur