missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Antibody (CL6913) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-76511AF700
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Spezifikation
| CD47 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| This antibody was developed using a recombinant protein derived from Q08722, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Immunology, Signal Transduction | |
| 961 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL6913 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur