missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD51/CD61 (Integrin alpha v beta 3) Monoclonal Antibody (23C6), eBioscience™
GREENER_CHOICE

Artikelnummer. 15267017
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
15267017 100 μg 100 Mikrogramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15267017

Marke: Invitrogen™ 14051982

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

Description: The 23C6 monoclonal antibody reacts with the human CD51/CD61 dimer, also known as the integrin alphav/beta3. CD51, an ~120 kDa surface molecule can also non-covalently associate with other beta subunits of the integrin family including beta1 (CD29), beta5 and beta6 to form receptors for extracellular matrix components. Heterodimers of CD51/CD61 are expressed by melanoma cells, endothelial cells and osteoclasts and at very low levels by platelets. The CD51/CD61 complex mediates adhesion to fibrinogen, fibronectin, vitronectin and thrombospondin. Applications Reported: The 23C6 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. 23C6 has also been reported in blocking of some adhesive processes. (Please use Functional Grade purified 23C6 in functional assays). Applications Tested: The 23C6 antibody has been tested by flow cytometric analysis of human melanoma cell line and peripheral blood. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufact...

CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CD51/CD61 (Integrin alpha v beta 3)
Anwendungen Flow Cytometry, Immunohistochemistry (Frozen)
Klassifikation Monoclonal
Klon 23C6
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.09% sodium azide; pH 7.2
Gen ITGAV
Gen-Zugriffsnummer P05106, P06756
Gen-Alias 1110004F14Rik; 2610028E01Rik; alpha v integrin; antigen CD61; antigen identified by monoclonal antibody L230; BDPLT16; BDPLT2; Cd51; CD61; D430040G12Rik; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; GT; INGRB3; integrin; integrin alpha V; integrin alpha-5 beta-3; integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; Integrin beta; integrin beta 3; integrin beta 3 (Cd61); Integrin beta3; integrin beta-3; integrin subunit alpha V; integrin subunit beta 3; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGAV; Itgb3; MSK8; platelet gpIIIa; platelet membrane glycoprotein IIIa; Vitronectin receptor; vitronectin receptor alpha polypeptide (VNRA); vitronectin receptor alpha subunit; vitronectin receptor subunit alpha; VNRA; VTNR
Gensymbole ITGAV, ITGB3
Wirtsspezies Mouse
Reinigungsverfahren Affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3685, 3690
Zielspezies Human
Inhalt und Lagerung 4°C
Produkttyp Antibody
Form Liquid
Isotype IgG1 κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.