Learn More
CD52 Antibody (Campath-1H) - Chimeric - Azide and BSA Free, Novus Biologicals™
Artikelnummer. 18240481 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.025 mg
0.2 mg
Packungsgröße:
0.20 Milligramm
25 Mikrogramm
Artikelnummer. Menge unitSize
18240481 0.025 mg 25 Mikrogramm
18094406 0.2 mg 0.20 Milligramm
2 options
Artikelnummer. 18240481
Marke: Novus Biologicals NBP2526450.025MG
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Monoclonal Antibody
CD52 Monoclonal specifically detects CD52 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, CyTOF-ready, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen CD52
Anwendungen Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon Campath-1H
Konjugat Unconjugated
Verdünnung Western Blot, Flow Cytometry 1:10 - 1:1000, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, CyTOF-ready
Zusammensetzung PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gen-Alias Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Gensymbole CD52
Wirtsspezies Rabbit
Immunogen Human lymphocytes.
Reinigungsverfahren Protein A or G purified
Menge 0.025 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 1043
Testspezifität Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.
Zielspezies Human, Rat
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG