Learn More
CD56 (NCAM) Mouse, anti-Human, PE, Clone: HCD56, STEMCELL Technologies
Artikelnummer. 17198531
Encompass_Preferred
Change view
Click to view available options
Menge:
100 Tests
25 Tests
Packungsgröße:
25 Tests
Artikelnummer. Menge unitSize
17198531 25 Tests 25 Tests
17188531 100 Tests -
2 options
Artikelnummer. 17198531
Marke: Stemcell Technologies 60021PE.1
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
  • The HCD56 antibody reacts with CD56 (NCAM), a member of the immunoglobulin (Ig) super family containing 5 Ig-like domains and two fibronectin type-3 domains in its extracellular portion
  • Multiple isoforms have been identified, including transmembrane and GPI-anchored forms
  • CD56 is subject to several types of post-translational modification, including addition of a polysialic acid moiety to the 5th Ig-like domain
  • The ∼140 kDa isoform of CD56 is considered the prototypic cell surface marker on human NK cells
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Chimerism Buffy Coat CD56 Positive Selection Kit (Catalog #17875) and EasySep™ Human CD56 Positive Selection Kit II (Catalog #17855); partial blocking may be observed, as well as EasySep™ Human CD2 Positive Selection Kit II (Catalog #17883) and EasySep™ Human NK Cell Enrichment Kit (Catalog #19055)
Spezifikation
Antigen CD56 (NCAM)
Anwendungen CyTOF, Flow Cytometry, Immunohistochemistry
Klassifikation Monoclonal
Klon HCD56
Konjugat PE
Wirtsspezies Mouse
Menge 25 Tests
Primär oder sekundär Primary
Zielspezies Human
Isotype IgG1 κ
Zelltyp NK Cells