missing translation for 'onlineSavingsMsg'
Learn More

CD57 Mouse anti-Human, FITC, Clone: NK-1, BD

Product Code. 15811459
Change view
Click to view available options
Menge:
100 Tests
25 Tests
Unit Size:
1 Stück
Product Code. Menge unitSize
15811459 25 Tests 1 Stück
15868308 100 Tests 1 Stück
2 options
This item is not returnable. View return policy

Product Code. 15811459

Brand: BD Biosciences 561906

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NK-1 monoclonal antibody specifically reacts with a 110 kDa carbohydrate antigen associated with myelin-associated glycoprotein expressed on 7-35% of normal peripheral blood lymphocytes including a subset of natural killer cells, a subset of CD8-positive peripheral blood T cells, and on some neural tissues. CD57 is not expressed on granulocytes, platelets, red blood cells or thymocytes. The function of CD57 is still unclear, however, its expression on T-cell subets occurs in late immune responses.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD57
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon NK-1
Konjugat FITC
Zusammensetzung Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Wirtsspezies Mouse
Reinigungsverfahren Affinity Purified
Menge 25 Tests
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.