missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Spezifikation
Spezifikation
| Antigen | CD63 |
| Anwendungen | Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 2585C |
| Konjugat | FITC |
| Verdünnung | Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready |
| Zusammensetzung | PBS |
| Gen-Alias | CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 |
| Wirtsspezies | Rabbit |
| Immunogen | Chinese Hamster Ovary cell line CHO-derived human CD63, Ala63-Val203, Accession # P08962-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?