Learn More
Invitrogen™ CD64 Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560080
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Medarex Patent Anti-Fc-Gamma-R1.
CD64 (FCGR1A) encodes a protein that plays an important role in the immune response. CD64 is a high-affinity Fc-gamma receptor and is one of three related gene family members located on chromosome 1. CD64 plays an important role in the immune response and its dysfunction has been studied in cervical adenitis and tetrasomy 21.
Spezifikation
| CD64 Biosimilar | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.