Learn More
Invitrogen™ CD7 Recombinant Rabbit Monoclonal Antibody (15), Invitrogen™
GREENER_CHOICE
Artikelnummer. 17759623
Change view
Click to view available options
Menge:
50 μL
100 μL
Packungsgröße:
100 Mikroliter
50 Mikroliter
Artikelnummer. Menge unitSize
17759623 100 μL 100 Mikroliter
17799873 50 μL 50 Mikroliter
2 options
Artikelnummer. 17759623
Marke: Invitrogen™ MA543071
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Recombinant Monoclonal Antibody
Specificity: Human CD7/LEU9
CD7, also known as gp40 or Leu9, is a 40 kDa receptor and member of the immunoglobulin gene superfamily. It features an N-terminal region (amino acids 1-107) that is highly homologous to Ig kappa light chains, while its carboxyl-terminal region is proline-rich, forming a stalk from which the Ig domain projects. CD7 is prominently expressed on the majority of immature and mature T lymphocytes, as well as T cell leukemias. It is also found on natural killer cells, a small subpopulation of normal B cells, and malignant B cells. CD7 plays a crucial role in modulating immune cell activity. Cross-linking of surface CD7 enhances T cell and NK cell functions, as evidenced by increased calcium flux, expression of adhesion molecules, cytokine secretion, and proliferation. CD7 directly associates with phosphoinositol 3-kinase, and its ligation induces the production of D-3 phosphoinositides and tyrosine phosphorylation. The expression of CD7 is an important marker in leukemia diagnostics, highlighting its significance in both normal immune function and disease states.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen CD7
Anwendungen ELISA, Immunohistochemistry (Paraffin)
Klassifikation Recombinant Monoclonal
Klon 15
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen CD7
Gen-Zugriffsnummer P09564
Gen-Alias Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41
Gensymbole CD7
Wirtsspezies Rabbit
Immunogen Recombinant Human CD7 protein.
Reinigungsverfahren Protein A
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 924
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG