Learn More
CD71 (Transferrin Receptor) Monoclonal Antibody (R17217 (RI7 217.1.4)), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
134.00 € - 374.00 €
Spezifikation
| Antigen | CD71 (Transferrin Receptor) |
|---|---|
| Klon | R17217 (RI7 217.1.4) |
| Konzentration | 0.2 mg/mL |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15341270
|
Invitrogen
46-0711-80 |
25 μg | |||||||
15351270
![]() |
Invitrogen™
46-0711-82 |
100 μg |
374.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Description: The R17217 monoclonal antibody reacts with mouse CD71, a 170-180 kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Expression level differences have been observed in different mouse strains. Applications Reported: This R17217 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R17217 (RI7 217.1.4) antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.Spezifikation
| CD71 (Transferrin Receptor) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q62351 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| R17217 (RI7 217.1.4) | |
| Flow Cytometry | |
| PerCP-eFluor 710 | |
| Rat | |
| Mouse | |
| 22042 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.