missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79A Antibody (706931) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals FAB69201F
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD79A Monoclonal antibody specifically detects CD79A in Human samples. It is validated for Flow Cytometry, CyTOF
Spezifikation
| CD79A | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 706931 | |
| Flow Cytometry, CyTOF-ready | |
| CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein | |
| Mouse myeloma cell line NS0-derived recombinant human CD79A, Leu33-Arg143, Accession # P11912 | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 973 | |
| Store at 4°C in the dark. | |
| IgG1 |
Berichtigung von Produktinhalten
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido