missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
257.00 € - 575.00 €
Spezifikation
| Antigen | CD8 |
|---|---|
| Konzentration | 0.2 mg/ml |
| Verdünnung | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18420062
|
Novus Biologicals
NBP2-29475-20ug |
20 ug |
257.00 €
20 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18749133
|
Novus Biologicals
NBP2-29475 |
0.1 mg |
575.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
CD8 Polyclonal specifically detects CD8 in Human, Mouse, Porcine samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Spezifikation
| CD8 | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Porcine | |
| P01732 | |
| 925 | |
| Recombinant fragment from human CD8 C-terminal cytoplasmic domain of alpha chain (exact sequence is proprietary). (Uniprot: P01732) | |
| Primary | |
| Store at 4C. | |
| 32 kDa |
| 0.2 mg/ml | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 10 mM PBS, pH 7.4 with 0.05% BSA with 0.05% Sodium Azide | |
| CD8, CD8a molecule, LEU2, p32 | |
| CD8A | |
| IgG | |
| Protein A purified | |
| CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts