missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD8 Antibody Pair [HRP]

Artikelnummer. 18665951 Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
15 Platten
5 Platten
Packungsgröße:
15 Stück
5 Stück
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18665951

Marke: Novus Biologicals™ NBP2793285Plates

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD8.
TRUSTED_SUSTAINABILITY

Specifications

Assay-Bereich 78.1 pg/mL
Assay-Empfindlichkeit: 78.13-5000 pg/mL
Assay Sandwich ELISA
Konjugat HRP
Produkttyp Antibody Pairs
Spezifität This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD8.
Zielspezies Human
Zur Verwendung mit (Anwendung) Sandwich ELISA
Gen-Alias CD8, CD8a molecule, LEU2, p32
Gen-ID (Entrez) 925
Gensymbol CD8A
Wirtsspezies Mouse
Kitinhalt
  • Standard: Each vial contains 180 ng of recombinant CD8.
  • Capture Antibody: 0.2 mg/mL of mouse anti-CD8 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL mouse anti-CD8 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Menge 5 Platten
    Kennzeichnung RUO
    Forschungsgebiet Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
    Lagerungsbedingungen Storage is content dependent.
    Ziel CD8
    Verfahren 1. Capture Antibody: Dilute to a working concentration of 0.5 ug/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.