Learn More
CD85k (Gp49b) Monoclonal Antibody (H1.1), Functional Grade, eBioscience™, Invitrogen™
Beschreibung
The MAb H1.1 recognizes gp49, a receptor with structural similarities to the PIR (paired Ig-activating receptors) and ILT/KIR(killer inhibitory receptors) family of proteins. Two closely linked genes on mouse chromosome 10 encode the related proteins gp49A and gp49B. These two receptors are almost 90% identical in their extracellular regions but differ in their cytoplasmic domains; gp49B contains an ITAM domain while gp49A does not and is extremely short. Expression of gp49 was originally identified on mast and LAK (IL-2 activated NK) cells. Expression on LAK cells has been shown to be predominantly gp49B with only a small amount of gp49A. Additionally gp49 is found on myeloid cells. Detailed function is not known but gp49B can inhibit cellular activation. The MAb H1.1 recognizes both gp49A and gp49B. The H1.1 antibody has been shown to have functional activity: when crosslinked along with anti-NK1.1 it can prevent cytokine release in KY-2 cells.
Spezifikation
Spezifikation
| Antigen | CD85k (Gp49b) |
| Anwendungen | Functional Assay, Neutralization |
| Klassifikation | Monoclonal |
| Klon | H1.1 |
| Konzentration | 1 mg/mL |
| Konjugat | Functional Grade |
| Zusammensetzung | PBS with no preservative; pH 7.2 |
| Gen | Lilrb4a |
| Gen-Zugriffsnummer | Q64281 |
| Gen-Alias | CD85 antigen-like family member K; CD85k; glycoprotein 49 B; glycoprotein 49b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; Mast cell surface glycoprotein Gp49B; monocyte inhibitory receptor HM18 |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.