Learn More
Invitrogen™ CD96 (TACTILE) Monoclonal Antibody (6A6), PE, eBioscience™

Beschreibung
Description: This 6A6 monoclonal antibody reacts with mouse CD96, a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gammadelta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity. This monoclonal antibody has been reported to block CD96 interaction with CD155 and Nectin-1. Applications Reported: This 6A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 6A6 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing fi...
Spezifikation
Spezifikation
| Antigen | CD96 (TACTILE) |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 6A6 |
| Konzentration | 0.2 mg/mL |
| Konjugat | PE |
| Zusammensetzung | PBS with 0.09% sodium azide; pH 7.2 |
| Gen | Cd96 |
| Gen-Zugriffsnummer | Q3U0X8 |
| Gen-Alias | 1700109I12Rik; Cd96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; Tactile; T-cell surface protein tactile |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.