Learn More
CD98/SLC3A2 Mouse anti-Human, Unconjugated, Clone: 2B10F5, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66883-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD98/SLC3A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD98/SLC3A2. This antibody recognizes human antigen. The CD98/SLC3A2 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Spezifikation
| CD98/SLC3A2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 2B10F5 | |
| Unconjugated | |
| 20 μL | |
| 6520 | |
| IgG2a |