Learn More
CDON Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17634-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.Spezifikation
| CDON | |
| Polyclonal | |
| Unconjugated | |
| CDON | |
| CDO, CDON, Cdon homolog (mouse), ORCAM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50937, 50938, 57810 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35158, Q32MD9, Q4KMG0 | |
| CDON | |
| CDON Fusion Protein Ag11904 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |