Beschreibung
CDON and BOC are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
Spezifikation
Spezifikation
| Antigen | CDON |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.25 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gen | CDON |
| Gen-Zugriffsnummer | O35158, Q32MD9, Q4KMG0 |
| Gen-Alias | CDO, CDON, Cdon homolog (mouse), ORCAM |
| Gensymbole | CDON |
| Mehr anzeigen |