missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM4 Monoclonal Antibody (568410)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561426
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
CEACAM4 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 4) is a Protein Coding gene. An important paralog of this gene is CEACAM3.
Spezifikation
| CEACAM4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75871 | |
| CEACAM4 | |
| E. coli-derived recombinant human CEACAM-4, Phe36-Val140. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 568410 | |
| Unconjugated | |
| CEACAM4 | |
| carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 10; carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; C-CAM4 protein; Ceacam10; CEACAM4; CEA-related cell adhesion molecule 10; CGM7; CGM7_HUMAN; NCA; Nonspecific cross-reacting antigen (NCA); non-specific cross-reacting antigen W236; nonspecific cross-reacting antigen W236 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1089 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur