Beschreibung
- For tubes, bottles, and plates in one system
- Versatile rotor portfolio with 15 rotors and many adapters to cover a wide range of vessel types and volumes
- Compatibility with existing Centrifuge 5810 R rotors to reuse rotor assets and ease the upgrade
- High performance with speeds up to 20,913 x g (14,000 rpm) for fast, reliable separations
- Refrigerated operation from -9°C to +40°C to protect temperature‐sensitive samples
- Sustainable R290 (propane) cooling with low global warming potential to support lab sustainability goals
- Intuitive control panel with program key and up to 35 user programs for quick, repeatable setup
- Low access height and Soft‐touch Lid for comfortable loading, unloading, and closing
- Eppendorf QuickLock™ rotor lids and caps for fast and safe rotor handling
- Quiet operation to support a focused and pleasant working environment
Spezifikation
Spezifikation
| Fassungsvermögen | 4 x 750 mL, 4 x 4 MTP |
| Max. RZB | 20,913 x g |
| Max. Geschwindigkeit | 14,000 rpm |
| Gekühlt | CFC free |
| Zur Verwendung mit (Geräte) | Tubes, Bottles, Plates |
| Rotor | None |
| Rotor inklusive | No |
| Gewährleistung | 2 Years |
| Abmessungen (L x B x H) außen | 70 x 64.6 x 35.3 cm |
| Nettogewicht | 89 kg/192.2 lb. |
| Mehr anzeigen |