missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CFHR4 Polyclonal Antibody

Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5149107
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene is a member of the complement factor H (CFH) gene family, and encodes one of the 5 CFH-related (CFHR) proteins. These 5 genes are closely linked to the CFH gene on chromosome 1q31-q32. The CFHRs are secreted plasma proteins synthesized primarily by the hepatocytes, and composed of highly-related short consensus repeats (SCRs). This protein enhances the cofactor activity of CFH, and is involved in complement regulation. It can associate with lipoproteins and may play a role in lipid metabolism. Alternatively spliced transcript variants encoding different isoforms (varying in the number of SCRs) have been described for this gene.
Spezifikation
| CFHR4 | |
| Polyclonal | |
| Unconjugated | |
| CFHR4 | |
| CFHL4; CFHR4; complement factor H; complement factor H related 4; complement factor H-related 4; complement factor H-related protein 4; FHR4; FHR-4 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 10877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q92496 | |
| CFHR4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Complement Factor H-related 4/CFHR4, Arg41-Glu331 (Ser79Leu). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur