missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The CHCHD1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Zelllinie | The lysate was created in HEK293T cells, using plasmid ID RC24571 and based on accession number NM_23298. The protein contains a C-terminal DDK tag. |
| Menge | 0,1 mg |
| Lagerungspuffer | RIPA buffer |
| Reinheit | Protein |
| Testspezifität | Homo sapiens coiled-coil-helix-coiled-coil-helix domain containing 1 (CHCHD1), mRNA. |
| Inhalt und Lagerung | -80°C. Avoid freeze-thaw cycles. |
| Produktlinie | Novus |
| Protein State | Native |
| Zur Verwendung mit (Anwendung) | Western Blot |
Nur für Forschungszwecke
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?