missing translation for 'onlineSavingsMsg'
Learn More

Chitinase 3-like 1/YKL-40 Antibody (CL12241) [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30287653 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
Product Code. Menge unitSize
30287653 0.1 mL 0.10 Milliliter
1 options
This item is not returnable. View return policy

Product Code. 30287653

Brand: Novus Biologicals NBP321212AF647

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chitinase 3-like 1/YKL-40 Monoclonal antibody specifically detects Chitinase 3-like 1/YKL-40 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chitinase 3-like 1/YKL-40
Anwendungen Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon CL12241
Konjugat Alexa Fluor 647
Zusammensetzung 50mM Sodium Borate
Gen-Alias AW208766, Brp39, Chi3l1, gp39, YKL-40
Wirtsspezies Mouse
Immunogen This antibody was developed using a synthetic peptide derived from P36222, with the exact immunogen sequence remaining proprietary.
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Cell Biology, Cytoskeleton Markers, Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 1116
Zielspezies Human
Inhalt und Lagerung Store at 4⪚C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.