Learn More
Chk2 Antibody (OTI5C4), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-71688MFV610
Artikelnummer. 18499636
  • 784.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Chk2 Monoclonal antibody specifically detects Chk2 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Chk2
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat, Canine
Purified
Flow Cytometry
OTI5C4
Flow Cytometry
CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog
Full length human recombinant protein of human CHEK2 (NP_009125) produced in HEK293T cell.
0.1 mL
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Stem Cell Markers
11200
Store at 4C in the dark.