missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cibenzolinsuccinat, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Cibenzoline Succinate |
| CAS | 100678-32-8 |
| Summenformel | C18 H18 N2 . C4 H6 O4 |
| Menge | 25 mg |
| Synonym | Butanedioic acid, compd. with 2-(2,2-diphenylcyclopropyl)-4,5-dihydro-1H-imidazole (1:1), 1H-Imidazole, 2-(2,2-diphenylcyclopropyl)-4,5-dihydro-, butanedioate (1:1) (9CI), Cibenol, Cibenzoline succinate, Cibenzoline succinate (1:1), Cifenline succinate, Cipralan, Exacor, Ro 22-7796/001 |
| InChI-Formel | InChI=1S/C18H18N2.C4H6O4/c1-3-7-14(8-4-1)18(15-9-5-2-6-10-15)13-16(18)17-19-11-12-20-17;5-3(6)1-2-4(7)8/h1-10,16H,11-13H2,(H,19,20);1-2H2,(H,5,6)(H,7,8) |
| SMILES | OC(=O)CCC(=O)O.C1CN=C(N1)C2CC2(c3ccccc3)c4ccccc4 |
| IUPAC-Name | Butandisäure; 2-(2,2-Diphenylcyclopropyl)-4,5-dihydro-1H-imidazol |
| Molekulargewicht (g/mol) | 380.44 |
| Formelmasse | 380.1736 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?