Learn More
cis-Bicyclo[3.3.0]octan-3,7-dion-3,7-bis(2',2'-dimethylpropyliden)diketal, TRC
Specifications
Specifications
| Chemischer Name oder Material | cis-Bicyclo[3.3.0]octane-3,7-dione-3,7-bis(2',2'-dimethylpropylidene) Diketal |
| CAS | 92007-38-0 |
| Summenformel | C18H30O4 |
| Menge | 250 mg |
| Synonym | cis-Tetrahydro-5,5,5'',5''-tetramethyldispiro[1,3-dioxane-2,2'(1'H)-pentalene-5'(3'H),2''-[1,3]dioxane] |
| InChI-Formel | InChI=1S/C18H30O4/c1-15(2)9-19-17(20-10-15)5-13-7-18(8-14(13)6-17)21-11-16(3,4)12-22-18/h13-14H,5-12H2,1-4H3 |
| SMILES | CC(CO1)(C)COC21CC3C(CC4(OCC(C)(C)CO4)C3)C2 |
| IUPAC-Name | 5,5,5'',5''-Tetramethyltetrahydro-1'H,3'H-dispiro[[1,3]dioxan-2,2'-pentalen-5',2''-[1,3]dioxan] |
| Molekulargewicht (g/mol) | 310.43 |
| Formelmasse | 310.21 |