missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cixutumumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558262
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Cixutumumab has been used in trials studying the treatment of Lung Cancer, Malignant Neoplasm, Leukemia, Mast-Cell, Non-Small-Cell Lung Carcinoma, and Adenocarcinoma of the Prostate. Cixutumumab is a highly specific recombinant human monoclonal antibody with high affinity for the insulin-like growth factor-I receptor (IGF-IR). IGF-IR is overexpressed in a variety of tumor types.
Spezifikation
| Cixutumumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IMC-A12; LY3012217 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur