Learn More
Invitrogen™ CK1 alpha Recombinant Rabbit Monoclonal Antibody (PSH04-46)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556414
Beschreibung
CK1 alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow, IP
CNSK1A1 (CK1 alpha 1) is a member of the casein kinase I (CKI) gene family, which encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric, ranging from 25 to 55 kD, and are found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. It can phosphorylate a large number of proteins. Participates in Wnt signaling. Phosphorylates CTNNB1 at 'Ser-45'. May phosphorylate PER1 and PER2. May play a role in segregating chromosomes during mitosis.
Spezifikation
| CK1 alpha | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P48729, P97633, Q8BK63 | |
| CSNK1A1 | |
| Recombinant protein within human CSNK1A1 aa 1-337/337. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| PSH04-46 | |
| Unconjugated | |
| CSNK1A1 | |
| 2610208K14Rik; 4632404G05Rik; 5430427P18Rik; casein kinase 1 alpha 1; casein kinase 1 alpha 1 like; casein kinase 1 alpha 1 S homeolog; casein kinase 1 alpha 1 S homeolog; casein kinase 1, alpha 1; casein kinase 1 alpha isoform; casein kinase 1, alpha 1; casein kinase 1alpha S; casein kinase 1-alphaL; casein kinase 1-alphaLS; casein kinase I; casein kinase I alpha; casein kinase I alpha L isoform; casein kinase I alpha LS; casein kinase I alpha S; Casein kinase I isoform alpha; casein kinase I isoform alpha; casein kinase I; casein kinase I-alpha; CHUNP6894; CK I alpha; CK1; ck1 alpha; CK1a; ck1alpha; CK1alphaS; CKI alpha; CK-I alpha; CKIa; CKI-alpha; clock regulator kinase; Csnk1a; csnk1a1; CSNK1A1 protein; csnk1a1.S; CSNK1A1/CK1; CSNK1A1L; down-regulated in lung cancer; epididymis secretory sperm binding protein Li 77p; HEL-S-77p; HLCDGP1; OTTHUMP00000224000; OTTHUMP00000224001; OTTHUMP00000224003; PRO2975; protein kinase; sb:eu866; wu:fb65a02; wu:fi30h04; wu:fj19c11; XELAEV_18021437mg; YIL035C; zgc:92158 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 113927, 1452, 93687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.