missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CKS2 Recombinant Rabbit Monoclonal Antibody (K01_3K53)

Artikelnummer. 30828101
Ansicht ändern
Click to view available options
Menge:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30828101 50 μL
30829995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30828101 Lieferant Invitrogen™ Lieferanten-Nr. MA565818
 Eingeschränkter Artikel
Verfügbar ab: 22-12-2026
Zum Warenkorb hinzufügen
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Recombinant Monoclonal Antibody

CKS2 Recombinant Monoclonal Antibody for Western Blot

CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifica

Antigen CKS2
Anwendungen Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_3K53
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gen Cks2
Gen-Zugriffsnummer P33552, P56390
Gen-Alias 1110038L14Rik; CDC28 protein kinase 2; CDC28 protein kinase regulatory subunit 2; CKS1(S. cerevisiae Cdc28/Cdc2 kinase subunit) homolog-2; CKS2; CKS-2; CKSHS2; Cyclin-dependent kinases regulatory subunit 2; RGD1562047; RP11-8C21.2
Gensymbole Cks2
Wirtsspezies Rabbit
Immunogen Peptide.
Reinigungsverfahren Protein A/G
Menge 50 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 1164, 498709, 66197
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.