missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-1 Antibody (CL3698) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61630V
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Claudin-1 Monoclonal antibody specifically detects Claudin-1 in Human samples. It is validated for Western Blot
Tekniske data
| Claudin-1 | |
| Monoclonal | |
| DyLight 405 | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| This antibody was developed using a recombinant protein derived from O95832, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Extracellular Matrix | |
| 9076 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL3698 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion