missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-18.2 Antibody (CLDN18.2/7300R), Alexa Fluor™ 594, Novus Biologicals™
Alle ansehen Bio Techne ProdukteDescription
Claudin-18.2 Monoclonal antibody specifically detects Claudin-18.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Claudin-18.2 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CLDN18.2/7300R |
| Konjugat | Alexa Fluor 594 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | claudin-18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fragment (around aa1-100) of human Claudin-18.2 protein (exact sequence is proprietary) |
| Reinigungsverfahren | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?