missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claudin 2 Monoclonal Antibody (12H12)
GREENER_CHOICE

Artikelnummer. 10392693
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
1 Stück
Artikelnummer. Menge unitSize
10392693 100 μg 1 Stück
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 10392693

Marke: Invitrogen™ 325600

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Mouse Monoclonal Antibody

This antibody reacts specifically with the ~22-23 kDa Claudin-2 protein. Reactivity has been confirmed with human, mouse, rat, and dog liver and kidney homogenates, as well MDCK and Caco-2 cell lysates, by western blotting and immunfluorescence. Reactivity has also been confirmed with formalin-fixed, paraffin-embedded (FFPE) human normal colon, spleen, and thyroid, and colon and breast cancer tissues by immunohistochemistry. For best results in immunohistochemistry (1-2 μg/mL) with formalin-fixed, paraffin-embedded (FFPE) tissues, heat induced epitope retrieval (HIER) with EDTA buffer, pH 8.0, is required prior to staining. In western blotting a non-specific band at 27 kDa is observed in some lysates. In Caco-2 cells this band appears stronger than the correct band at 22 kDa. An alternative product, rabbit anti-Claudin-2 (Cat. No. 516100) may be used for western blotting of these lysates.

Tight junctions are specialized regions of cell-cell contact that are particularly abundant in luminal epithelial cell sheets. In freeze-fracture electron micrographs, tight junctions are visualized as belt-like bands of anastomosing sealing strands (TJ strands) that completely encircle the lateral surfaces of each cell. TJ strands on adjacent cells are presumed to interact with each other to form a sort of 'molecular gasket' that prevents ions, water and other molecules from leaking between cells and thus, from one side of the sheet to the other. In addition to this so-called 'barrier' function, the 'fence' function of tight junctions plays an important role in maintaining epithelial cell-polarity by blocking the diffusion of membrane proteins between apical (luminal) and basolateral cell surfaces. Confinement of, for example, the glucose symport to apical surfaces allows glucose to be transported vectorially from the lumen, through the cell, and into the bloodstream. Several peripheral membrane proteins are associated with tight junctions including ZO-1, ZO-2, ZO-3 (members of membrane-associated guanylate-kinase family), cingulin, the 7H6 antigen, Rab-3b, symplekin. While their precise functions are not known, roles for these proteins have been suggested in tight junction assembly and maintenance; signal transduction; and the regulation of tight junction permeability. Furthermore, a growing body of evidence suggests that actin filaments play a major role in regulating tight junction permeability. Until recently, the only transmembrane protein known to be associated with tight junctions was occludin, an ~65 kDa protein with four transmembrane domains. Despite widespread expectation, a critical structural role for occludin in TJ strands was ruled out by the observation of apparently normal tight junctions formed between cells disrupted at both occludin alleles. Fortunately, a closer examination of isolated tight junctions uncovered two related ~22 kDa, four-transmembrane domain proteins, claudin-1 and claudin-2, with no similarity to occludin. In contrast to occludin, which induces only a small number of short strands at cell-cell contact sites when introduced into fibroblasts lacking tight junctions, claudin-1 and -2 induce networks of strands characteristic of true tight junctions. Though inconclusive, these findings suggest that claudin-1 and -2 are major structural components of TJ strands and that occludin plays some other accessory role. Excitement in the tight junction field continues to rise following the recent discovery of claudins -3, -4, -5, -6, -7, and -8 and experiments suggesting that tight junctions in different tissues are comprised of different sets of claudin family proteins. Claudin-1 and claudin-2 connect to the actin cytoskeleton through ZO-1; Claudin-2 functions as a paracellular channel with cation (Na+) selectivity at tight junctions. The expression of claudin-2 is restricted to the liver and kidney, with small amounts also found in the brain.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Claudin 2
Anwendungen Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassifikation Monoclonal
Klon 12H12
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.1% sodium azide; pH 7.4
Gen CLDN2
Gen-Zugriffsnummer O88552, P57739, Q95KM6
Gen-Alias AL022813; claudin 2; Claudin2; claudin-2; CLD2; Cldn2; integral membrane protein claudin-2; PSEC0059; RGD1560247; SP82; UNQ705/PRO1356
Gensymbole CLDN2
Wirtsspezies Mouse
Immunogen Synthetic peptide corresponding to a 26 amino acid sequence at the C-terminus of mouse Claudin-2.
Reinigungsverfahren Protein A
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 12738, 300920, 403649, 9075
Zielspezies Human, Mouse, Rat, Canine
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG2b
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.