missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 3 Monoclonal Antibody (CL13128)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA546538
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 74%.
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
Spezifikation
| Claudin 3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15551 | |
| CLDN3 | |
| Synthetic Peptide. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL13128 | |
| Unconjugated | |
| CLDN3 | |
| AI182374; C7orf1; claudin 3; claudin-3; CLDN3; clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-R2; CPE-receptor 2; CPETR2; hRVP1; integral membrane protein claudin-3; mRVP1; Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; RVP1; tight junction protein; ventral prostate.1 protein; ventral prostate.1 protein homolog; ventral prostate.1-like protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 1365 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur