missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 4 Recombinant Mouse Monoclonal Antibody (8G7)

Mouse Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA550226
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Claudin 4 Recombinant Monoclonal Antibody for ELISA
Claudin-4 belongs to the 20-member family of claudins, transmembrane cellular adhesion proteins localized at epithelial and endothelial tight junctions. Claudins demonstrate distinct, tissue-specific expression patterns, and are essential components of tight junction charge-specific channels that regulate paracellular ion flux. Claudin-4 is a component of tight junction strands and may play a role in internal organ development and function during pre- and postnatal life. The overexpression of Claudin-4 has been shown to decrease paracellular electrical conductance due to a selective decrease in Na+ permeability, with no significant decrease in Cl- permeability. In ovarian carcinoma cells, Claudin-3 and Claudin-4 exhibit upregulation at both mRNA and protein levels. Claudin-4 is also overexpressed in pancreatic cancer tissues and cell lines, and in other epithelial cancers. The Claudin-4 gene is deleted in Williams-Beuren syndrome, a neurodevelopmental disorder affecting multiple systems.
Spezifikation
| Claudin 4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| O14493 | |
| Cldn4 | |
| Recombinant Human CLDN4 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry | |
| 8G7 | |
| Unconjugated | |
| Cldn4 | |
| Cep-r; claudin 4; CLAUDIN4; claudin-4; CLDN4; Clostridium perfringens enterotoxin receptor; Clostridium perfringens enterotoxin receptor 1; CPER; CPE-R; CPE-receptor; Cpetr; CPETR1; h CPE-receptor; hCPE-R; WBSCR8; Williams-Beuren syndrome chromosomal region 8 protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1364 | |
| -20°C or -80°C if preferred | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur