missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claudin 8 Recombinant Superclonal™ Antibody (4HCLC)

Artikelnummer. 15387254
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
15387254 100 μg 100 Mikrogramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 15387254

Marke: Invitrogen™ 710222

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with equine, mouse and rat based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.

Claudin-8 belongs to the claudin family of integral membrane proteins. Claudins are responsible for the formation of tight junction strands, and are connected with the actin cytoskeleton mediated by ZO-1. Claudin-8 is a tight junction protein expressed in several distal nephron segments, suggesting that it may have a role as a paracellular cation barrier protein. Claudin-8 is expressed in the inner ear, and the umbrella cell tight junctions in the bladder epithelium.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Claudin 8
Anwendungen Western Blot, Immunocytochemistry
Klassifikation Recombinant Superclonal
Klon 4HCLC
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.09% sodium azide
Gen CLDN8
Gen-Zugriffsnummer P56748
Gen-Alias AI648025; claudin 8; claudin-8; CLDN8; epididymis secretory protein Li 79; HEL-S-79; UNQ779/PRO1573
Gensymbole CLDN8
Wirtsspezies Rabbit
Immunogen Peptide corresponding to amino acids 206-225 of human claudin-8.
Reinigungsverfahren Protein A
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 9073
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.