Learn More
Invitrogen™ Claudin 9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5104427
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total Claudin 9.
CLDN9 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
Spezifikation
| Claudin 9 | |
| Polyclonal | |
| Unconjugated | |
| Cldn9 | |
| claudin 9; claudin-9; CLDN9; nmf329 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287099, 56863, 9080 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95484, Q9Z0S7 | |
| Cldn9 | |
| A synthesized peptide derived from human CLDN9(Accession O95484), corresponding to amino acid residues G179-V217. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |