Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Y Union, Animal-Derivative-Free Polypropylene
Alle ansehen Cole-Parmer Produkte
Click to view available options
Innendurchmesser (angloamerikanisch):
1/16 in.
1/2 in.
1/4 in.
1/8 in.
1 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Packungsgröße:
10 Stück
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS |
| Innendurchmesser (angloamerikanisch) | 3/32 in. |
| Min. Betriebstemperatur (metrisch) | -23°C |
| Verpackungsmenge | 10/Pk. |
| Auslassanschluss | Hose Barb Y |
| Sterilisationsmethode | 121°C |
| Form | Y |
| Farbe | Natural |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.