Learn More
Cole-Parmer Cleanroom Hose Barb to Plug Fittings, Straight Adapter, Natural Kynar™
Alle ansehen Cole-Parmer Produkte
Click to view available options
Schlauchanschluss:
1/16 in. Schlauchanschluss
1/2 in. Schlauchanschluss
1/4 in. Schlauchanschluss
1/8 in. Schlauchanschluss
1 in. Schlauchanschluss
5/32 in. Schlauchanschluss
Packungsgröße:
10 Stück
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Spezifikation
Spezifikation
| Zertifizierung/Konformität | USP;Non-DEHP;Non-Phthalate |
| Min. Betriebstemperatur (metrisch) | -35°C |
| Verpackungsmenge | 10/Pk. |
| Schlauchanschluss | 5/32 in. Schlauchanschluss |
| Auslassanschluss | Hose Barb x Plug |
| Sterilisationsmethode | E-beam/gamma irradiation, EtO, or autoclave up to 135°C |
| Farbe | Natural |