missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor X Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marke: Novus Biologicals NBP1-33320
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Coagulation Factor X Polyclonal specifically detects Coagulation Factor X in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| Coagulation Factor X | |
| Polyclonal | |
| Unconjugated | |
| 1XPBS, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
| F10 | |
| Recombinant protein encompassing a sequence within the center region of human Factor X. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2159 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.96mg/mL | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| coagulation factor X, EC 3.4.21, EC 3.4.21.6, factor Xa, FX, FXA, prothrombinase, Stuart factor, Stuart-Prower factor | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur