Learn More
Invitrogen™ Cofilin Recombinant Rabbit Monoclonal Antibody (24GB4775)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557363
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cofilin Recombinant Monoclonal Antibody for Western Blot, Flow
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.
Spezifikation
| Cofilin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CFL1 | |
| 18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1072, 12631, 29271 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB4775 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18760, P23528, P45592 | |
| CFL1 | |
| A synthesized peptide derived from human Cofilin (aa 116-129). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |