Learn More
Collagen IV Antibody (rCOL4/4742), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP3-08487AF350
Artikelnummer. 18591184
  • ONLINE EXCLUSIVE PRICE
    643.00 €
    607.95 € / 100 Mikroliter
    Sparen 35.05 €
    5% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Collagen IV Monoclonal antibody specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Collagen IV
Monoclonal
Alexa Fluor 350
BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR
Purified human placental extract
100 μg
Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
1282
Store at 4°C in the dark.
IgG2a κ
Immunohistochemistry (Paraffin)
rCOL4/4742
50mM Sodium Borate
Mouse
Protein A or G purified