missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Colon Matched Primary Tumor and Metastatic Tumor Slides - Frozen

Artikelnummer. 18618950 Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
10 Objektträger
Packungsgröße:
10 Stück
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18618950

Marke: Novus Biologicals™ NBP277663

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Tissue Slides

TRUSTED_SUSTAINABILITY

Specifications

Menge 10 Objektträger
Zur Verwendung mit (Anwendung) Immunohistochemistry (IHC), Immunohistochemistry (Frozen), ISH, Hematoxylin and Eosin Stain
Spezies Human
Zielspezifität Colon
Inhalt und Lagerung Store at -70°C. Avoid freeze-thaw cycles.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.