Learn More
Invitrogen™ Complement C5a Monoclonal Antibody (C17/5)

Mouse Monoclonal Antibody
Marke: Invitrogen™ GAU0250502
Beschreibung
Specificity: GAU 025-05 reacts with human C5a/C5a (desArg) and cross-reacts with baboon C5a/C5a (desArg). Reactivity: For ELISA applications, recombinant C5a coated onto a microtiter well results in a strong reaction. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
The protein encoded by this gene is the fifth component of complement, which plays an important role in inflammatory and cell killing processes. This protein is comprised of alpha and beta polypeptide chains that are linked by a disulfide bridge. An activation peptide, C5a, which is an anaphylatoxin that possesses potent spasmogenic and chemotactic activity, is derived from the alpha polypeptide via cleavage with a convertase. The C5b macromolecular cleavage product can form a complex with the C6 complement component, and this complex is the basis for formation of the membrane attack complex, which includes additional complement components. Mutations in this gene cause complement component 5 deficiency, a disease where patients show a propensity for severe recurrent infections. Defects in this gene have also been linked to a susceptibility to liver fibrosis and to rheumatoid arthritis.
Spezifikation
| Complement C5a | |
| Monoclonal | |
| 1.05 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P01031 | |
| C5 | |
| Purified human C5a. | |
| 400 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| C17/5 | |
| Unconjugated | |
| C5 | |
| anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905 | |
| Mouse | |
| Protein A | |
| RUO | |
| 727 | |
| 4°C, store in dark | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.