missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA529093
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Recommended positive controls: THP-1, BCL-1, human plasma. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C5b-9 membrane attack complexes are assembled from five precursor molecules in the serum. Proteolytic cleavage of C5 by C5 convertase generates C5b which initiates assembly of the C5b-9 complex. The last step of C5b-9 complex formation involves polymerization of C9 which accompanies insertion of the complex into the cell membrane. During formation of C5b-8 and C9 polymerization, neoantigens are generated which are unique to the C5b-9 complex and are not present on any of the individual native complex components. The complement regulatory proteins CD59 and complement S-protein can both prevent C5b-9 insertion into the cell membrane. The formed SC5b-9 complex is unable to attach to cells and is cytolytically inactive. C5b-9 is involved in the progression of chronic proteinuric renal disease by mediating continuous tubulointerstitial damage. Early tubulointerstitial injury in the remnant kidney can be improved when C5b-9 complex forming is abrogated.
Spezifikation
| Complement C9 | |
| Polyclonal | |
| Unconjugated | |
| C9 | |
| ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117512, 12279, 735 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.62 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P02748, P06683, Q62930 | |
| C9 | |
| Recombinant protein encompassing a sequence within the center region of human C9. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur