Learn More
Novus Biologicals™ Complement Component C2 Antibody (ARGX-117) - Humanized, Novus Biologicals™
Artikelnummer. 30045951 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
1 mg
100 μg
50 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
50 Mikrogramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30045951 50 μg 50 Mikrogramm
30046802 1 mg 1 Milligramm
30046068 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30045951 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP32821550ug
Please to purchase this item. Need a web account? Register with us today!
Human Monoclonal Antibody
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Complement Component C2
Anwendungen Flow Cytometry, ELISA, Functional Assay
Klassifikation Monoclonal
Klon ARGX-117
Konjugat Unconjugated
Zusammensetzung Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gen-Zugriffsnummer P06681
Gen-Alias C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43
Wirtsspezies Human
Immunogen Complement C2
Reinigungsverfahren Protein A purified
Menge 50 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 717
Rekonstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Zielspezies Human
Inhalt und Lagerung Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1