Learn More
Tuttnauer Conductivity Probe
Alle ansehen Tuttnauer Produkte
Click to view available options
:
Conductivity Probe; For use with: Tiva Washer
Beschreibung
- Conductivity Probe Accessory
- Checks the water purity level at the end of rinsing.
- The machine repeats the rinse cycle again to remove any residue if the conductivity probe detects a water purity level, expressed in μS/cm, higher than the preset one.
Spezifikation
Spezifikation
| Material | Plastic |
| Produkttyp | Probe |
| Zur Verwendung mit (Geräte) | Tiva Washer |
| Menge | 1 Ea. |